Assay Details
| Gene Symbol: SETD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555949.2 | XP_005556006.1 | 5 - 6 | 1669 | 81 | |
| XM_005555950.2 | XP_005556007.1 | 5 - 6 | 1669 | 81 | ||
| XM_015450850.1 | XP_015306336.1 | 5 - 6 | 1669 | 81 | ||
More Information
Set Membership: |
|

SETD7
Genomic Map