Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285354.1 | NP_001272283.1 | 3 - 4 | 1077 | 72 | |
| XM_005557474.2 | XP_005557531.1 | 3 - 4 | 1051 | 72 | ||
| XM_005557475.2 | XP_005557532.1 | 3 - 4 | 974 | 72 | ||
| XM_005557476.2 | XP_005557533.1 | 3 - 4 | 820 | 72 | ||
| XM_015451390.1 | XP_015306876.1 | 2 - 3 | 669 | 72 | ||
| GenBank mRNA | AB168797.1 | - | 3 - 4 | 1077 | 72 | |
More Information
Set Membership: |
|

NUDT12
Genomic Map