Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575329.2 | XP_005575386.2 | 1 - 2 | 579 | 109 | |
| XM_015433229.1 | XP_015288715.1 | 1 - 2 | 581 | 109 | ||
More Information
Set Membership: |
|

ANKRD23
Genomic Map