Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542866.2 | XP_005542923.1 | 9 - 10 | 1243 | 130 | |
| XM_005542867.2 | XP_005542924.1 | 9 - 10 | 1174 | 130 | ||
| XM_015433424.1 | XP_015288910.1 | 10 - 11 | 1369 | 130 | ||
| GenBank mRNA | AB070141.1 | - | 9 - 10 | 1195 | 130 | |
More Information
Set Membership: |
|

DNAI3
Genomic Map