Assay Details
| Gene Symbol: FANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283847.1 | NP_001270776.1 | 2 - 3 | 279 | 81 | |
| XM_005566712.2 | XP_005566769.1 | 2 - 3 | 293 | 81 | ||
| XM_015455895.1 | XP_015311381.1 | 2 - 3 | 293 | 81 | ||
| XM_015455897.1 | XP_015311383.1 | 2 - 3 | 219 | 81 | ||
| GenBank mRNA | AB179439.1 | - | 2 - 3 | 279 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015455897.1 transcripts.
Set Membership: |
|

FANK1
Genomic Map