Assay Details
| Gene Symbol: LOC102133073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581113.2 | XP_005581170.1 | 3 - 4 | 3107 | 107 | |
| XM_015436869.1 | XP_015292355.1 | 11 - 12 | 3942 | 107 | ||
| XM_015436872.1 | XP_015292358.1 | 3 - 4 | 2825 | 107 | ||
More Information
Set Membership: |
|

LOC102133073
Genomic Map