Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592828.2 | XP_005592885.1 | 7 - 8 | 1358 | 72 | |
| XM_015442959.1 | XP_015298445.1 | 7 - 8 | 1719 | 72 | ||
| GenBank mRNA | AB052145.1 | - | 7 - 8 | 1082 | 72 | |
More Information
Set Membership: |
|

ZNF276
Genomic Map