Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572258.2 | XP_005572315.2 | 1305 | 125 | ||
| GenBank mRNA | AB168691.1 | - | 1311 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC63| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572258.2 | XP_005572315.2 | 1305 | 125 | ||
| GenBank mRNA | AB168691.1 | - | 1311 | 125 | ||
Set Membership: |
|