Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556932.2 | XP_005556989.1 | 3 - 4 | 975 | 137 | |
| XM_005556934.1 | XP_005556991.1 | 3 - 4 | 975 | 137 | ||
| XM_005556935.2 | XP_005556992.1 | 3 - 4 | 884 | 137 | ||
More Information
Set Membership: |
|

SETD9
Genomic Map