Assay Details
| Gene Symbol: SLCO6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557442.2 | XP_005557499.1 | 4 - 5 | 1089 | 89 | |
| XM_005557443.2 | XP_005557500.1 | 4 - 5 | 1089 | 89 | ||
| GenBank mRNA | AB072739.1 | - | 4 - 5 | 737 | 89 | |
More Information
Set Membership: |
|

SLCO6A1
Genomic Map