Assay Details
| Gene Symbol: ETFBKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570500.2 | XP_005570557.1 | 2 - 3 | 590 | 90 | |
| XM_015430596.1 | XP_015286082.1 | 2 - 3 | 840 | 90 | ||
| XM_015430598.1 | XP_015286084.1 | 2 - 3 | 534 | 90 | ||
More Information
Set Membership: |
|

ETFBKMT
Genomic Map