Assay Details
| Gene Symbol: UGT3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556722.2 | XP_005556779.1 | 2 - 3 | 320 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT3A2| Gene Symbol: UGT3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556722.2 | XP_005556779.1 | 2 - 3 | 320 | 127 | |
Set Membership: |
|