Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575727.2 | XP_005575784.1 | 4 - 5 | 1364 | 153 | |
| XM_005575728.2 | XP_005575785.1 | 4 - 5 | 970 | 153 | ||
| XM_015433585.1 | XP_015289071.1 | 5 - 6 | 1473 | 153 | ||
More Information
Set Membership: |
|

SPRED2
Genomic Map