Assay Details
| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566351.2 | XP_005566408.1 | 4 - 5 | 2430 | 154 | |
| XM_015456645.1 | XP_015312131.1 | 4 - 5 | 2569 | 154 | ||
| XM_015456646.1 | XP_015312132.1 | 4 - 5 | 2455 | 154 | ||
| XM_015456647.1 | XP_015312133.1 | 4 - 5 | 2500 | 154 | ||
More Information
Set Membership: |
|

CALHM3
Genomic Map