Assay Details
| Gene Symbol: STIMATE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547385.2 | XP_005547442.1 | 2 - 3 | 335 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STIMATE| Gene Symbol: STIMATE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547385.2 | XP_005547442.1 | 2 - 3 | 335 | 97 | |
Set Membership: |
|