Assay Details
| Gene Symbol: KLHL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581624.2 | XP_005581681.1 | 1 | 434 | 120 | |
| XM_015437103.1 | XP_015292589.1 | 1 | 434 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005581624.2, XM_015437103.1 transcripts.
Set Membership: |
|

KLHL9
Genomic Map