Assay Details
| Gene Symbol: GPR50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594829.2 | XP_005594886.1 | 1 - 2 | 329 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR50| Gene Symbol: GPR50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594829.2 | XP_005594886.1 | 1 - 2 | 329 | 103 | |
Set Membership: |
|