Assay Details
| Gene Symbol: FOXA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589636.2 | XP_005589693.1 | 1 - 2 | 271 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXA3| Gene Symbol: FOXA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589636.2 | XP_005589693.1 | 1 - 2 | 271 | 96 | |
Set Membership: |
|