Assay Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588851.2 | XP_005588908.1 | 2 | 2136 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FFAR3| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588851.2 | XP_005588908.1 | 2 | 2136 | 113 | |
Set Membership: |
|