Assay Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541255.2 | XP_005541312.1 | 1 - 2 | 271 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005541255.2 transcripts.
Set Membership: |
|

NHLH1
Genomic Map