Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592303.2 | XP_005592360.1 | 1 - 2 | 153 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005592303.2 transcripts.
Set Membership: |
|

PSKH1
Genomic Map