Assay Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549058.2 | XP_005549115.1 | 2 - 3 | 568 | 80 | |
| XM_015447050.1 | XP_015302536.1 | 2 - 3 | 568 | 80 | ||
| XM_015447051.1 | XP_015302537.1 | 2 - 3 | 604 | 80 | ||
| XM_015447052.1 | XP_015302538.1 | 3 - 4 | 335 | 80 | ||
| XM_015447053.1 | XP_015302539.1 | 1 - 2 | 365 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015447051.1, XM_015447052.1, XM_015447053.1 transcripts.
Set Membership: |
|

RBAK
Genomic Map