Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592685.2 | XP_005592742.1 | 8 - 9 | 1289 | 103 | |
| XM_015443185.1 | XP_015298671.1 | 7 - 8 | 985 | 103 | ||
| XM_015443186.1 | XP_015298672.1 | 7 - 8 | 1008 | 103 | ||
| XM_015443188.1 | XP_015298674.1 | 7 - 8 | 870 | 103 | ||
| XM_015443189.1 | XP_015298675.1 | 7 - 8 | 870 | 103 | ||
| XM_015443190.1 | XP_015298676.1 | 7 - 8 | 870 | 103 | ||
More Information
Set Membership: |
|

ZDHHC7
Genomic Map