Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590501.2 | XP_005590558.1 | 3 | 2647 | 142 | |
| XM_005590502.2 | XP_005590559.1 | 3 | 2972 | 142 | ||
| XM_015441559.1 | XP_015297045.1 | 3 | 2440 | 142 | ||
More Information
Set Membership: |
|

ZNF460
Genomic Map