Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550565.2 | XP_005550622.1 | 5 | 2856 | 124 | |
| XM_015447963.1 | XP_015303449.1 | 5 | 2853 | 124 | ||
More Information
Set Membership: |
|

MDFIC
Genomic Map