Assay Details
| Gene Symbol: LOC102118812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569123.2 | XP_005569180.2 | 2 | 5702 | 76 | |
| GenBank mRNA | AY781390.1 | - | 3 | 1531 | 76 | |
More Information
Set Membership: |
|

LOC102118812
Genomic Map