Assay Details
| Gene Symbol: CXCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319397.1 | NP_001306326.1 | 1 | 984 | 79 | |
| XM_005546891.2 | XP_005546948.1 | 2 | 2919 | 79 | ||
| XM_005546892.2 | XP_005546949.1 | 2 | 1377 | 79 | ||
| XM_005546893.2 | XP_005546950.1 | 2 | 1070 | 79 | ||
| XM_005546894.2 | XP_005546951.1 | 2 | 1212 | 79 | ||
| GenBank mRNA | AF291671.1 | - | 1 | 984 | 79 | |
More Information
Set Membership: |
|

CXCR6
Genomic Map