Assay Details
| Gene Symbol: FOSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577214.2 | XP_005577271.1 | 3 - 4 | 1021 | 72 | |
| XM_015434437.1 | XP_015289923.1 | 3 - 4 | 995 | 72 | ||
More Information
Set Membership: |
|

FOSL1
Genomic Map