Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567363.2 | XP_005567420.1 | 3 | 3221 | 77 | |
| XM_015457230.1 | XP_015312716.1 | 4 | 8253 | 77 | ||
| XM_015457231.1 | XP_015312717.1 | 2 | 5091 | 77 | ||
| XM_015457232.1 | XP_015312718.1 | 2 | 3020 | 77 | ||
More Information
Set Membership: |
|

ELFN2
Genomic Map