Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562511.2 | XP_005562568.1 | 2 - 3 | 837 | 69 | |
| XM_005562512.2 | XP_005562569.1 | 3 - 4 | 882 | 69 | ||
| XM_005562514.2 | XP_005562571.1 | 2 - 3 | 820 | 69 | ||
| XM_005562515.2 | XP_005562572.1 | 2 - 3 | 764 | 69 | ||
| XM_015454423.1 | XP_015309909.1 | 4 - 5 | 994 | 69 | ||
More Information
Set Membership: |
|

CLN8
Genomic Map