Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569905.2 | XP_005569962.1 | 1 - 2 | 282 | 67 | |
| XM_015430277.1 | XP_015285763.1 | 1 - 2 | 252 | 67 | ||
More Information
Set Membership: |
|

TAPBPL
Genomic Map