Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589756.2 | XP_005589813.1 | 4 - 5 | 631 | 56 | |
| XM_005589757.2 | XP_005589814.1 | 4 - 5 | 443 | 56 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map