Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562829.2 | XP_005562886.2 | 3 - 4 | 422 | 73 | |
| XM_015454576.1 | XP_015310062.1 | 3 - 4 | 443 | 73 | ||
| XM_015454578.1 | XP_015310064.1 | 3 - 4 | 421 | 73 | ||
| XM_015454579.1 | XP_015310065.1 | 3 - 4 | 378 | 73 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map