Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583199.2 | XP_005583256.1 | 1 - 2 | 162 | 76 | |
| XM_005583200.2 | XP_005583257.1 | 2 - 3 | 566 | 76 | ||
| XM_005583201.2 | XP_005583258.1 | 2 - 3 | 214 | 76 | ||
More Information
Set Membership: |
|

LYRM9
Genomic Map