Assay Details
| Gene Symbol: TNFAIP8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587579.2 | XP_005587636.1 | 1 - 2 | 133 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005587579.2 transcripts.
Set Membership: |
|

TNFAIP8L1
Genomic Map