Assay Details
| Gene Symbol: HTR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548396.2 | XP_005548453.1 | 4 | 1773 | 100 | |
| XM_005548398.2 | XP_005548455.1 | 4 | 1911 | 100 | ||
| XM_005548399.2 | XP_005548456.1 | 3 | 1667 | 100 | ||
More Information
Set Membership: |
|

HTR1F
Genomic Map