Assay Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585651.2 | XP_005585708.1 | 2 | 696 | 109 | |
| XM_015439154.1 | XP_015294640.1 | 2 | 1054 | 109 | ||
| XM_015439157.1 | XP_015294643.1 | 2 | 876 | 109 | ||
More Information
Set Membership: |
|

SERTM1
Genomic Map