Assay Details
| Gene Symbol: LOC102125550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583750.1 | XP_005583807.1 | 2 - 3 | 352 | 58 | |
| XM_005583751.2 | XP_005583808.1 | 2 - 3 | 277 | 58 | ||
More Information
Set Membership: |
|

LOC102125550
Genomic Map