Assay Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583814.2 | XP_005583871.1 | 4 | 5863 | 78 | |
| XM_015438347.1 | XP_015293833.1 | 4 | 5303 | 78 | ||
More Information
Set Membership: |
|

SRSF1
Genomic Map