Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551204.2 | XP_005551261.1 | 1 - 2 | 1797 | 89 | |
| XM_005551205.2 | XP_005551262.1 | 1 - 2 | 2057 | 89 | ||
More Information
Set Membership: |
|

CHPF2
Genomic Map