Assay Details
| Gene Symbol: DEDD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541214.2 | XP_005541271.1 | 5 - 6 | 1000 | 73 | |
| XM_005541215.2 | XP_005541272.1 | 5 - 6 | 777 | 73 | ||
| XM_015454663.1 | XP_015310149.1 | 5 - 6 | 1863 | 73 | ||
| XM_015454668.1 | XP_015310154.1 | 5 - 6 | 1729 | 73 | ||
| XM_015454669.1 | XP_015310155.1 | 5 - 6 | 825 | 73 | ||
| XM_015454670.1 | XP_015310156.1 | 4 - 5 | 2016 | 73 | ||
| XM_015454675.1 | XP_015310161.1 | 5 - 6 | 1858 | 73 | ||
| XM_015454681.1 | XP_015310167.1 | 5 - 6 | 779 | 73 | ||
| XM_015454691.1 | XP_015310177.1 | 5 - 6 | 748 | 73 | ||
More Information
Set Membership: |
|

DEDD
Genomic Map