Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563663.2 | XP_005563720.1 | 4 - 5 | 782 | 88 | |
| XM_005563664.2 | XP_005563721.1 | 4 - 5 | 1048 | 88 | ||
| XM_005563665.2 | XP_005563722.1 | 4 - 5 | 1145 | 88 | ||
| XM_005563666.2 | XP_005563723.1 | 4 - 5 | 736 | 88 | ||
| XM_015455009.1 | XP_015310495.1 | 3 - 4 | 1389 | 88 | ||
More Information
Set Membership: |
|

OSGIN2
Genomic Map