Assay Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547131.2 | XP_005547188.1 | 2 - 3 | 715 | 98 | |
| XM_005547132.2 | XP_005547189.1 | 3 - 4 | 744 | 98 | ||
| XM_005547133.2 | XP_005547190.1 | 3 - 4 | 749 | 98 | ||
| XM_005547134.2 | XP_005547191.1 | 2 - 3 | 540 | 98 | ||
| XM_005547135.2 | XP_005547192.1 | 2 - 3 | 833 | 98 | ||
| XM_005547136.2 | XP_005547193.1 | 3 - 4 | 555 | 98 | ||
| XM_005547137.1 | XP_005547194.1 | 2 - 3 | 775 | 98 | ||
More Information
Set Membership: |
|

DAG1
Genomic Map