Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578309.2 | XP_005578366.1 | 4 - 5 | 460 | 119 | |
| XM_005578310.2 | XP_005578367.1 | 4 - 5 | 460 | 119 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map