Assay Details
| Gene Symbol: KCNMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558502.2 | XP_005558559.1 | 3 - 4 | 1037 | 96 | |
| XM_005558503.2 | XP_005558560.1 | 3 - 4 | 512 | 96 | ||
More Information
Set Membership: |
|

KCNMB1
Genomic Map