Assay Details
| Gene Symbol: CREBZF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579246.2 | XP_005579303.1 | 1 | 1000 | 138 | |
| XM_005579247.2 | XP_005579304.1 | 1 | 1000 | 138 | ||
| XM_005579249.2 | XP_005579306.1 | 1 | 2020 | 138 | ||
| XM_015436076.1 | XP_015291562.1 | 1 | 2018 | 138 | ||
| XM_015436077.1 | XP_015291563.1 | 1 | 1000 | 138 | ||
| XM_015436078.1 | XP_015291564.1 | 1 | 1000 | 138 | ||
More Information
Set Membership: |
|

CREBZF
Genomic Map