Assay Details
| Gene Symbol: LPAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319516.1 | NP_001306445.1 | 3 - 4 | 1183 | 85 | |
| XM_015440872.1 | XP_015296358.1 | 2 - 3 | 1786 | 85 | ||
| XM_015440874.1 | XP_015296360.1 | 3 - 4 | 1827 | 85 | ||
| GenBank mRNA | AB060872.1 | - | 3 - 4 | 1183 | 85 | |
More Information
Set Membership: |
|

LPAR2
Genomic Map