Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558119.2 | XP_005558176.1 | 3 - 4 | 1508 | 69 | |
| XM_005558120.2 | XP_005558177.1 | 3 - 4 | 1465 | 69 | ||
| XM_005558121.2 | XP_005558178.1 | 2 - 3 | 2108 | 69 | ||
| XM_005558123.2 | XP_005558180.1 | 2 - 3 | 2169 | 69 | ||
| XM_005558124.2 | XP_005558181.1 | 2 - 3 | 1354 | 69 | ||
| XM_005558125.2 | XP_005558182.1 | 2 - 3 | 1311 | 69 | ||
| XM_015452075.1 | XP_015307561.1 | 2 - 3 | 1662 | 69 | ||
More Information
Set Membership: |
|

KCTD16
Genomic Map