Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542698.2 | XP_005542755.1 | 4 - 5 | 540 | 106 | |
| XM_005542699.2 | XP_005542756.1 | 3 - 4 | 916 | 106 | ||
More Information
Set Membership: |
|

TMED5
Genomic Map