Assay Details
| Gene Symbol: LOC102129196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593214.2 | XP_005593271.1 | 5 | 777 | 86 | |
| XM_005593215.2 | XP_005593272.1 | 2 | 466 | 86 | ||
| XM_015443651.1 | XP_015299137.1 | 5 | 606 | 86 | ||
| XM_015443652.1 | XP_015299138.1 | 3 | 616 | 86 | ||
More Information
Set Membership: |
|

LOC102129196
Genomic Map